2-{[(4-Acetylphenyl)amino]carbonyl}cyclohexanecarboxylic acid

Supplier: Matrix Scientific

030147-500MG
101829-572EA 109.25 USD
101829-572
2-{[(4-Acetylphenyl)amino]carbonyl}cyclohexanecarboxylic acid
2-{[(4-Acetylphenyl)amino]carbonyl}cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉NO₄
MDL Number: MFCD00608378
CAS Number: 324061-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR