6-{[(4-Acetylphenyl)amino]carbonyl}cyclohex-3-ene-1-carboxylic acid

Supplier: Matrix Scientific

030146-500MG
101829-570EA 109.25 USD
101829-570
6-{[(4-Acetylphenyl)amino]carbonyl}cyclohex-3-ene-1-carboxylic acid
6-{[(4-Acetylphenyl)amino]carbonyl}cyclohex-3-ene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₇NO₄
MDL Number: MFCD02090930
CAS Number: 445260-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR