5-[(4-Acetylphenyl)amino]-5-oxopentanoic acid

Supplier: Matrix Scientific

030145-500MG
101829-568EA 221.54 USD
101829-568
5-[(4-Acetylphenyl)amino]-5-oxopentanoic acid
5-[(4-Acetylphenyl)amino]-5-oxopentanoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD00033293
CAS Number: 174885-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR