2-(4-Acetylphenoxy)acetamide

Supplier: Matrix Scientific

030142-500MG
101829-562EA 146.15 USD
101829-562
2-(4-Acetylphenoxy)acetamide
2-(4-Acetylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD01922552
CAS Number: 42017-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR