1-(5-Bromothien-2-yl)butan-1-one

Supplier: Matrix Scientific

030141-500MG
101829-560EA 146.15 USD
101829-560
1-(5-Bromothien-2-yl)butan-1-one
1-(5-Bromothien-2-yl)butan-1-one

Hazard: Irritant

Formula: C₈H₉BrOS
MDL Number: MFCD06657522
CAS Number: 106652-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR