1-(4-Isopropoxyphenyl)pentan-1-one

Supplier: Matrix Scientific

030138-500MG
101829-554EA 109.25 USD
101829-554
1-(4-Isopropoxyphenyl)pentan-1-one
1-(4-Isopropoxyphenyl)pentan-1-one

Hazard: Irritant

Formula: C₁₄H₂₀O₂
MDL Number: MFCD02375373
CAS Number: 524732-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR