1-[4-(2-Piperidin-1-ylethoxy)phenyl]propan-1-one

Supplier: Matrix Scientific

030133-500MG
101829-544EA 146.15 USD
101829-544
1-[4-(2-Piperidin-1-ylethoxy)phenyl]propan-1-one
1-[4-(2-Piperidin-1-ylethoxy)phenyl]propan-1-one

Hazard: Irritant

Formula: C₁₆H₂₃NO₂
MDL Number: MFCD02609610
CAS Number: 101255-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR