Ethyl (5-bromo-2,3-dioxo-2,3-dihydro-1H-indol-1-yl)acetate

Supplier: Matrix Scientific

030129-500MG
101829-536EA 221.54 USD
101829-536
Ethyl (5-bromo-2,3-dioxo-2,3-dihydro-1H-indol-1-yl)acetate
Ethyl (5-bromo-2,3-dioxo-2,3-dihydro-1H-indol-1-yl)acetate

Hazard: Irritant

Formula: C₁₂H₁₀BrNO₄
MDL Number: MFCD04128671
CAS Number: 754235-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR