N-(3-Acetylphenyl)methanesulfonamide

Supplier: Matrix Scientific

030127-500MG
101829-532EA 109.25 USD
101829-532
N-(3-Acetylphenyl)methanesulfonamide
N-(3-Acetylphenyl)methanesulfonamide

Hazard: Irritant

Formula: C₉H₁₁NO₃S
MDL Number: MFCD00664903
CAS Number: 2417-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR