1-[4-(2-Morpholin-4-ylethoxy)phenyl]propan-1-one

Supplier: Matrix Scientific

030125-500MG
101829-528EA 193.85 USD
101829-528
1-[4-(2-Morpholin-4-ylethoxy)phenyl]propan-1-one
1-[4-(2-Morpholin-4-ylethoxy)phenyl]propan-1-one

Hazard: Irritant

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD01695064
CAS Number: 2089-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR