7-Ethyl-1H-indole-2,3-dione

Supplier: Matrix Scientific

Synonyms: 7-Ethylisatin

030124-500MG
101829-526EA 267.69 USD
101829-526
7-Ethyl-1H-indole-2,3-dione
7-Ethyl-1H-indole-2,3-dione

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00047222
CAS Number: 79183-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR