5-Methoxyisatin
Supplier: Matrix Scientific
Synonyms:
5-Methoxy-1H-indole-2,3-dione
, 5-Methoxyindoline-2,3-dione
030122-100G
030122-25G
101829-522EA
704.62
USD
101829-522
101829-520
5-Methoxyisatin
5-Methoxyisatin
Hazard: Irritant
Formula:
C₉H₇NO₃ MW: 177.16 g/mol |
MDL Number:
MFCD00169023 CAS Number: 39755-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...