5,7-Dimethylisatin

Supplier: Matrix Scientific

Synonyms: 5,7-Dimethyl-1H-indole-2,3-dione5,7-Dimethylindoline-2,3-dione

030119-500MG
101829-510EA 193.85 USD
101829-510
5,7-Dimethylisatin
5,7-Dimethylisatin

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00047219
CAS Number: 39603-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR