N-(3-Acetylphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

030116-500MG
101829-504EA 146.15 USD
101829-504
N-(3-Acetylphenyl)cyclobutanecarboxamide
N-(3-Acetylphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD02176923
CAS Number: 109920-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR