N-(4-Acetylphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

030115-500MG
101829-502EA 146.15 USD
101829-502
N-(4-Acetylphenyl)cyclobutanecarboxamide
N-(4-Acetylphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD02176900
CAS Number: 423745-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR