N-(3-Acetylphenyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

030114-500MG
101829-500EA 146.15 USD
101829-500
N-(3-Acetylphenyl)cyclopentanecarboxamide
N-(3-Acetylphenyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₁₄H₁₇NO₂
MDL Number: MFCD03362685
CAS Number: 540757-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR