N-(3-Acetylphenyl)-2-ethylbutanamide

Supplier: Matrix Scientific

030112-500MG
101829-496EA 146.15 USD
101829-496
N-(3-Acetylphenyl)-2-ethylbutanamide
N-(3-Acetylphenyl)-2-ethylbutanamide

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD02090928
CAS Number: 540521-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR