N-(3-Acetylphenyl)-2-methylpropanamide

Supplier: Matrix Scientific

030110-500MG
101829-492EA 109.25 USD
101829-492
N-(3-Acetylphenyl)-2-methylpropanamide
N-(3-Acetylphenyl)-2-methylpropanamide

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD02090927
CAS Number: 80945-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR