N-(4-Acetylphenyl)-2-methylpropanamide
Supplier: Matrix Scientific
030109-500MG
101829-490EA
146.15
USD
101829-490
N-(4-Acetylphenyl)-2-methylpropanamide
N-(4-Acetylphenyl)-2-methylpropanamide
Hazard: Irritant
Formula:
C₁₂H₁₅NO₂ |
MDL Number:
MFCD00448088 CAS Number: 128184-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...