N-(4-Acetylphenyl)-2,2-dimethylpropanamide
Supplier: Matrix Scientific
030107-500MG
101829-486EA
146.15
USD
101829-486
N-(4-Acetylphenyl)-2,2-dimethylpropanamide
N-(4-Acetylphenyl)-2,2-dimethylpropanamide
Hazard: Irritant
Formula:
C₁₃H₁₇NO₂ |
MDL Number:
MFCD00584423 CAS Number: 130599-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...