N-(4-Acetylphenyl)-2-(2,4-dichlorophenoxy)acetamide

Supplier: Matrix Scientific

030103-500MG
101829-478EA 109.25 USD
101829-478
N-(4-Acetylphenyl)-2-(2,4-dichlorophenoxy)acetamide
N-(4-Acetylphenyl)-2-(2,4-dichlorophenoxy)acetamide

Hazard: Irritant

Formula: C₁₆H₁₃Cl₂NO₃
MDL Number: MFCD00028562
CAS Number: 139194-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR