N-(3-Acetylphenyl)-2-(4-chlorophenoxy)acetamide

Supplier: Matrix Scientific

030102-500MG
101829-476EA 109.25 USD
101829-476
N-(3-Acetylphenyl)-2-(4-chlorophenoxy)acetamide
N-(3-Acetylphenyl)-2-(4-chlorophenoxy)acetamide

Hazard: Irritant

Formula: C₁₆H₁₄ClNO₃
MDL Number: MFCD00708013
CAS Number: 303795-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR