N-(3-Acetylphenyl)-2-(4-chlorophenyl)acetamide

Supplier: Matrix Scientific

030098-500MG
101829-468EA 109.25 USD
101829-468
N-(3-Acetylphenyl)-2-(4-chlorophenyl)acetamide
N-(3-Acetylphenyl)-2-(4-chlorophenyl)acetamide

Hazard: Irritant

Formula: C₁₆H₁₄ClNO₂
MDL Number: MFCD01353447
CAS Number: 72116-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR