N-(4-Acetylphenyl)-2-(4-methoxyphenyl)acetamide

Supplier: Matrix Scientific

030095-500MG
101829-462EA 109.25 USD
101829-462
N-(4-Acetylphenyl)-2-(4-methoxyphenyl)acetamide
N-(4-Acetylphenyl)-2-(4-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₇H₁₇NO₃
MDL Number: MFCD00859446
CAS Number: 314766-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR