N-(4-Acetylphenyl)-2-phenylacetamide

Supplier: Matrix Scientific

030093-500MG
101829-458EA 109.25 USD
101829-458
N-(4-Acetylphenyl)-2-phenylacetamide
N-(4-Acetylphenyl)-2-phenylacetamide

Hazard: Irritant

Formula: C₁₆H₁₅NO₂
MDL Number: MFCD00431008
CAS Number: 89246-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR