N-(3-Acetylphenyl)-4-(4-chloro-2-methylphenoxy)butanamide

Supplier: Matrix Scientific

030088-500MG
101829-448EA 146.15 USD
101829-448
N-(3-Acetylphenyl)-4-(4-chloro-2-methylphenoxy)butanamide
N-(3-Acetylphenyl)-4-(4-chloro-2-methylphenoxy)butanamide

Hazard: Irritant

Formula: C₁₉H₂₀ClNO₃
MW: 345.83 g/mol
MDL Number: MFCD01115605
CAS Number: 353467-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR