N-(4-Acetylphenyl)-4-(4-chloro-2-methylphenoxy)butanamide
Supplier: Matrix Scientific
030087-500MG
101829-446EA
146.15
USD
101829-446
N-(4-Acetylphenyl)-4-(4-chloro-2-methylphenoxy)butanamide
N-(4-Acetylphenyl)-4-(4-chloro-2-methylphenoxy)butanamide
Hazard: Irritant
Formula:
C₁₉H₂₀ClNO₃ MW: 345.83 g/mol |
MDL Number:
MFCD01115603 CAS Number: 353467-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...