N-(3-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)acetamide
Supplier: Matrix Scientific
030086-500MG
101829-444EA
146.15
USD
101829-444
N-(3-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)acetamide
N-(3-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)acetamide
Hazard: Irritant
Formula:
C₁₇H₁₆ClNO₃ MW: 317.77 g/mol |
MDL Number:
MFCD01115557 CAS Number: 314023-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...