N-(4-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)acetamide

Supplier: Matrix Scientific

030085-500MG
101829-442EA 109.25 USD
101829-442
N-(4-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)acetamide
N-(4-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₇H₁₆ClNO₃
MDL Number: MFCD01025443
CAS Number: 282104-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR