N-(4-Acetylphenyl)-4-methoxybenzamide

Supplier: Matrix Scientific

030083-500MG
101829-438EA 146.15 USD
101829-438
N-(4-Acetylphenyl)-4-methoxybenzamide
N-(4-Acetylphenyl)-4-methoxybenzamide

Hazard: Irritant

Formula: C₁₆H₁₅NO₃
MDL Number: MFCD00573199
CAS Number: 72269-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR