N-(3-Acetylphenyl)-4-tert-butylbenzamide

Supplier: Matrix Scientific

030080-500MG
101829-432EA 109.25 USD
101829-432
N-(3-Acetylphenyl)-4-tert-butylbenzamide
N-(3-Acetylphenyl)-4-tert-butylbenzamide

Hazard: Irritant

Formula: C₁₉H₂₁NO₂
MDL Number: MFCD00703784
CAS Number: 303790-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR