N-(4-Acetylphenyl)-4-tert-butylbenzamide

Supplier: Matrix Scientific

030079-500MG
101829-430EA 146.15 USD
101829-430
N-(4-Acetylphenyl)-4-tert-butylbenzamide
N-(4-Acetylphenyl)-4-tert-butylbenzamide

Hazard: Irritant

Formula: C₁₉H₂₁NO₂
MDL Number: MFCD01035360
CAS Number: 301157-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR