N-(3-Acetylphenyl)-2-methylbenzamide

Supplier: Matrix Scientific

030078-500MG
101829-428EA 146.15 USD
101829-428
N-(3-Acetylphenyl)-2-methylbenzamide
N-(3-Acetylphenyl)-2-methylbenzamide

Hazard: Irritant

Formula: C₁₆H₁₅NO₂
MDL Number: MFCD00433394
CAS Number: 315669-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR