N-(3-Acetylphenyl)-3-methylbenzamide

Supplier: Matrix Scientific

030076-500MG
101829-424EA 146.15 USD
101829-424
N-(3-Acetylphenyl)-3-methylbenzamide
N-(3-Acetylphenyl)-3-methylbenzamide

Hazard: Irritant

Formula: C₁₆H₁₅NO₂
MDL Number: MFCD01047336
CAS Number: 315669-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR