N-(3-Acetylphenyl)-4-methylbenzamide

Supplier: Matrix Scientific

030074-500MG
101829-420EA 109.25 USD
101829-420
N-(3-Acetylphenyl)-4-methylbenzamide
N-(3-Acetylphenyl)-4-methylbenzamide

Hazard: Irritant

Formula: C₁₆H₁₅NO₂
MDL Number: MFCD00433318
CAS Number: 303793-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR