N-(4-Acetylphenyl)-4-methylbenzamide

Supplier: Matrix Scientific

030073-500MG
101829-418EA 146.15 USD
101829-418
N-(4-Acetylphenyl)-4-methylbenzamide
N-(4-Acetylphenyl)-4-methylbenzamide

Hazard: Irritant

Formula: C₁₆H₁₅NO₂
MDL Number: MFCD00433240
CAS Number: 72269-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR