N-(3-Acetylphenyl)-3-methylthiophene-2-carboxamide

Supplier: Matrix Scientific

030072-500MG
101829-416EA 146.15 USD
101829-416
N-(3-Acetylphenyl)-3-methylthiophene-2-carboxamide
N-(3-Acetylphenyl)-3-methylthiophene-2-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₃NO₂S
MDL Number: MFCD03758983
CAS Number: 515846-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR