N-(4-Acetylphenyl)-5-chlorothiophene-2-carboxamide

Supplier: Matrix Scientific

030067-500MG
101829-406EA 146.15 USD
101829-406
N-(4-Acetylphenyl)-5-chlorothiophene-2-carboxamide
N-(4-Acetylphenyl)-5-chlorothiophene-2-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₂S
MDL Number: MFCD04406399
CAS Number: 391225-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR