N-(4-Acetylphenyl)-5-methyl-2-furamide

Supplier: Matrix Scientific

030065-500MG
101829-402EA 109.25 USD
101829-402
N-(4-Acetylphenyl)-5-methyl-2-furamide
N-(4-Acetylphenyl)-5-methyl-2-furamide

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD01346042
CAS Number: 549540-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR