N-(3-Acetylphenyl)-2-fluorobenzamide

Supplier: Matrix Scientific

030064-500MG
101829-400EA 146.15 USD
101829-400
N-(3-Acetylphenyl)-2-fluorobenzamide
N-(3-Acetylphenyl)-2-fluorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₂FNO₂
MDL Number: MFCD00745331
CAS Number: 303091-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR