N-(3-Acetylphenyl)-3-fluorobenzamide

Supplier: Matrix Scientific

030062-500MG
101829-396EA 146.15 USD
101829-396
N-(3-Acetylphenyl)-3-fluorobenzamide
N-(3-Acetylphenyl)-3-fluorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₂FNO₂
MDL Number: MFCD00091764
CAS Number: 194783-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR