N-(4-Acetylphenyl)-3-fluorobenzamide

Supplier: Matrix Scientific

030061-500MG
101829-394EA 146.15 USD
101829-394
N-(4-Acetylphenyl)-3-fluorobenzamide
N-(4-Acetylphenyl)-3-fluorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₂FNO₂
MDL Number: MFCD00431052
CAS Number: 312703-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR