N-(4-Acetylphenyl)-4-fluorobenzamide

Supplier: Matrix Scientific

030059-500MG
101829-390EA 109.25 USD
101829-390
N-(4-Acetylphenyl)-4-fluorobenzamide
N-(4-Acetylphenyl)-4-fluorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₂FNO₂
MDL Number: MFCD00430093
CAS Number: 300668-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR