N-(3-Acetylphenyl)-3-chlorobenzamide

Supplier: Matrix Scientific

030056-500MG
101829-384EA 146.15 USD
101829-384
N-(3-Acetylphenyl)-3-chlorobenzamide
N-(3-Acetylphenyl)-3-chlorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₂ClNO₂
MDL Number: MFCD00095786
CAS Number: 196408-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR