1-(4-Isopropoxyphenyl)propan-1-one

Supplier: Matrix Scientific

030051-500MG
101829-374EA 116.92 USD
101829-374
1-(4-Isopropoxyphenyl)propan-1-one
1-(4-Isopropoxyphenyl)propan-1-one

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD02176649
CAS Number: 35081-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR