1-(4-Propoxyphenyl)propan-1-one

Supplier: Matrix Scientific

030050-500MG
101829-372EA 116.92 USD
101829-372
1-(4-Propoxyphenyl)propan-1-one
1-(4-Propoxyphenyl)propan-1-one

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD01694976
CAS Number: 5736-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR