1-(4-Ethoxyphenyl)propan-1-one

Supplier: Matrix Scientific

030049-500MG
101829-370EA 116.92 USD
101829-370
1-(4-Ethoxyphenyl)propan-1-one
1-(4-Ethoxyphenyl)propan-1-one

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01010688
CAS Number: 35031-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR