1-[4-(Allyloxy)phenyl]ethanone

Supplier: Matrix Scientific

030047-500MG
101829-366EA 116.92 USD
101829-366
1-[4-(Allyloxy)phenyl]ethanone
1-[4-(Allyloxy)phenyl]ethanone

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD01673002
CAS Number: 2079-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR