N-(3-Acetylphenyl)-2-methyl-3-furamide

Supplier: Matrix Scientific

030045-500MG
101829-362EA 146.15 USD
101829-362
N-(3-Acetylphenyl)-2-methyl-3-furamide
N-(3-Acetylphenyl)-2-methyl-3-furamide

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD01834745
CAS Number: 352704-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR