N-(4-Acetylphenyl)-2-methyl-3-furamide
Supplier: Matrix Scientific
030044-500MG
101829-360EA
146.15
USD
101829-360
N-(4-Acetylphenyl)-2-methyl-3-furamide
N-(4-Acetylphenyl)-2-methyl-3-furamide
Hazard: Irritant
Formula:
C₁₄H₁₃NO₃ |
MDL Number:
MFCD01356015 CAS Number: 424807-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...